missing translation for 'onlineSavingsMsg'
Learn More

N-methyl-(1H-benzimidazol-5-ylmethyl)amine dioxalate, 90%, Thermo Scientific™

Product Code. 10654405
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 910036-85-0
Molecular Formula C13H15N3O8
Molecular Weight (g/mol) 341.276
MDL Number MFCD09025842
InChI Key HDQRBNFLWRHBAA-UHFFFAOYSA-N
Synonym n-methyl-1h-benzimidazol-5-ylmethyl amine dioxalate, 1h-1,3-benzodiazol-5-ylmethyl methyl amine; bis oxalic acid, n-methyl-1h-benzo d imidazol-5-ylmethyl amine dioxalate, oxalic acid-1-1h-benzimidazol-6-yl-n-methylmethanamine 2/1, 3h-1,3-benzodiazol-5-ylmethyl methyl amine; bis oxalic acid
PubChem CID 18525786
IUPAC Name 1-(3H-benzimidazol-5-yl)-N-methylmethanamine;oxalic acid
SMILES CNCC1=CC2=C(C=C1)N=CN2.C(=O)(C(=O)O)O.C(=O)(C(=O)O)O

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 341.27
Percent Purity 90%
Chemical Name or Material N-methyl-(1H-benzimidazol-5-ylmethyl)amine dioxalate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.