missing translation for 'onlineSavingsMsg'
Learn More
N-Methyl-3-(3-methyl-1,2,4-oxadiazol-5-yl)benzylamine, 95%, Thermo Scientific™ Product Code.: 10215894

N-Methyl-3-(3-methyl-1,2,4-oxadiazol-5-yl)benzylamine, 95%, Thermo Scientific™

Product Code. 10215894
1 g, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10215894

Brand: Thermo Scientific Maybridge CC49546DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 921938-59-2
Molecular Formula C11H13N3O
Molecular Weight (g/mol) 203.245
MDL Number MFCD09817473
InChI Key FWMVARCHFJQGJP-UHFFFAOYSA-N
Synonym n-methyl-3-3-methyl-1,2,4-oxadiazol-5-yl benzylamine, methyl 3-3-methyl-1,2,4-oxadiazol-5-yl phenyl methyl amine, n-methyl-1-3-3-methyl-1,2,4-oxadiazol-5-yl phenyl methanamine, 3-methyl-5-3-methylamino methyl phenyl-1,2,4-oxadiazole, methyl 3-3-methyl 1,2,4-oxadiazol-5-yl phenyl methyl amine
PubChem CID 24229501
IUPAC Name N-methyl-1-[3-(3-methyl-1,2,4-oxadiazol-5-yl)phenyl]methanamine
SMILES CC1=NOC(=N1)C2=CC(=CC=C2)CNC

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 203.24
Percent Purity 95%
Chemical Name or Material N-methyl-3-(3-methyl-1, 2, 4-oxadiazol-5-yl)benzylamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Methyl-3-(3-methyl-1,2,4-oxadiazol-5-yl)benzylamine, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.