missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-3-methylbenzamide, 97%, Thermo Scientific™

Product Code. 15452758
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74786-81-5
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.193
MDL Number MFCD01211937
InChI Key WKSODUAHKWBHQZ-UHFFFAOYSA-N
Synonym n-methyl-3-methylbenzamide, 3,n-dimethylbenzamide, benzamide,n,3-dimethyl, benzamide, n,3-dimethyl-9ci, n-methyl 3-methylphenyl carboxamide
PubChem CID 966002
IUPAC Name N,3-dimethylbenzamide
SMILES CC1=CC=CC(=C1)C(=O)NC

Specifications

Quantity 250 mg
Formula Weight 149.18
Percent Purity 97%
Chemical Name or Material N-Methyl-3-methylbenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'