missing translation for 'onlineSavingsMsg'
Learn More

n-methyl-[5-(morpholinomethyl)thien-2-ylmethyl]amine, 97%, Thermo Scientific™

Product Code. 10123034
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10123034

Brand: Thermo Scientific Maybridge CC73446DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 893742-71-7
Molecular Formula C11H18N2OS
Molecular Weight (g/mol) 226.34
MDL Number MFCD06803277
InChI Key LJPRPFHZPNCEIQ-UHFFFAOYSA-N
Synonym n-methyl-5-morpholinomethyl thien-2-ylmethyl amine, methyl 5-morpholin-4-ylmethyl thiophen-2-yl methyl amine, n-methyl-1-5-morpholin-4-ylmethyl thiophen-2-yl methylamine, 2-methylamino methyl-5-morpholin-4-ylmethyl thiophene, 2-thiophenemethanamine,n-methyl-5-4-morpholinylmethyl, methyl 5-morpholin-4-ylmethyl 2-thienyl methyl amine, n-methyl-1-5-morpholinomethyl thiophen-2-yl methanamine, n-methyl-1-5-morpholin-4-yl methyl thiophen-2-yl methanamine
PubChem CID 20098901
SMILES CNCC1=CC=C(CN2CCOCC2)S1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name N-methyl-1-[5-(morpholin-4-ylmethyl)thiophen-2-yl]methanamine
Formula Weight 226.34
Percent Purity 97%
Chemical Name or Material N-methyl-[5-(morpholinomethyl)thien-2-ylmethyl]amine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.