Learn More
n-Methyl-(chroman-6-ylmethyl)amine, 97%, Thermo Scientific™
119.00 € - 295.00 €
Chemical Identifiers
CAS | 950603-17-5 |
---|---|
Molecular Formula | C11H15NO |
Molecular Weight (g/mol) | 177.247 |
MDL Number | MFCD12198110 |
InChI Key | OYZVOCNCDRSGDZ-UHFFFAOYSA-N |
Synonym | n-methyl-chroman-6-ylmethyl amine, 1-chroman-6-yl-n-methylmethanamine, 3,4-dihydro-2h-1-benzopyran-6-ylmethyl methyl amine, 1-3,4-dihydro-2h-1-benzopyran-6-yl-n-methylmethanamine, chroman-6-ylmethyl methylamine, 1-3,4-dihydro-2h-chromen-6-yl-n-methyl-methanamine |
PubChem CID | 43811035 |
IUPAC Name | 1-(3,4-dihydro-2H-chromen-6-yl)-N-methylmethanamine |
SMILES | CNCC1=CC2=C(C=C1)OCCC2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10632495
|
Thermo Scientific Maybridge
CC32846CB |
250 mg | Amber Glass Bottle |
119.00 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10642495
|
Thermo Scientific Maybridge
CC32846DA |
1 g | Amber Glass Bottle |
295.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
950603-17-5 | |
177.247 | |
OYZVOCNCDRSGDZ-UHFFFAOYSA-N | |
43811035 | |
CNCC1=CC2=C(C=C1)OCCC2 |
C11H15NO | |
MFCD12198110 | |
n-methyl-chroman-6-ylmethyl amine, 1-chroman-6-yl-n-methylmethanamine, 3,4-dihydro-2h-1-benzopyran-6-ylmethyl methyl amine, 1-3,4-dihydro-2h-1-benzopyran-6-yl-n-methylmethanamine, chroman-6-ylmethyl methylamine, 1-3,4-dihydro-2h-chromen-6-yl-n-methyl-methanamine | |
1-(3,4-dihydro-2H-chromen-6-yl)-N-methylmethanamine |
Specifications
950603-17-5 | |
100.0 | |
C11H15NO | |
n-methyl-chroman-6-ylmethyl amine, 1-chroman-6-yl-n-methylmethanamine, 3,4-dihydro-2h-1-benzopyran-6-ylmethyl methyl amine, 1-3,4-dihydro-2h-1-benzopyran-6-yl-n-methylmethanamine, chroman-6-ylmethyl methylamine, 1-3,4-dihydro-2h-chromen-6-yl-n-methyl-methanamine | |
CNCC1=CC2=C(C=C1)OCCC2 | |
177.247 | |
177.25 | |
N-methyl-(chroman-6-ylmethyl)amine |
96.5 | |
97% | |
MFCD12198110 | |
OYZVOCNCDRSGDZ-UHFFFAOYSA-N | |
1-(3,4-dihydro-2H-chromen-6-yl)-N-methylmethanamine | |
43811035 | |
97% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
Your input is important to us. Please complete this form to provide feedback related to the content on this product.