missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Methyl-D-glucamine, 99%

Product Code. 10783881
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

N-Methyl-D-glucamine, CAS # 6284-40-8, also known as meglumine, is a base compound commonly used in combination with iodinated organic compounds in contrast medium.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Information


• N-Methyl-D-glucamine is a hexosamine compound that is frequently used in combination with iodinated organic compounds in contrast media such as iothalamate meglumine, iodipamide meglumine, and diatrizoate meglumine


Applications


• It is also useful when applied in ion-exchange resins to chelate boron ions selectively
• It is considered a base and frequently used in the preparation of salts of certain acids
• It is also applied in the synthesis of surface-active agents and laboratory dyes

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6284-40-8
Molecular Formula C7H17NO5
Molecular Weight (g/mol) 195.21
MDL Number MFCD00004707
InChI Key MBBZMMPHUWSWHV-BDVNFPICSA-N
Synonym meglumine, n-methyl-d-glucamine, n-methylglucamine, meglumin, 1-deoxy-1-methylamino-d-glucitol, methylglucamin, meglumina, 1-deoxy-1-methylaminosorbitol, d-glucitol, 1-deoxy-1-methylamino, n-methyl-d--glucamine
PubChem CID 8567
ChEBI CHEBI:59732
IUPAC Name (2R,3R,4R,5S)-6-(methylamino)hexane-1,2,3,4,5-pentol
SMILES CNCC(C(C(C(CO)O)O)O)O

Specifications

Absorbance 0.03 max. (20 w/v% aq. soln.) at 420nm
CAS Min % 98.5
CAS Max % 100.0
Melting Point 128.0°C to 132.0°C
Color White
Loss on Drying 1% max. (1 g, 100°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 100 g
Beilstein 04, IV, 1914
Merck Index 15, 6150
Specific Rotation Condition − 16.50 (20.00°C c=10,H2O,on dry sub)
Specific Rotation − 16.50
Solubility Information Solubility in water: 100 g/100 mL water (25°C). Other solubilities: 1.2 g/100 mL alcohol (25°C), 21 g/100 mL alcohol (70°C)
Formula Weight 195.21
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material N-Methyl-D-glucamine, 99%
Show More Show Less
Product Identifier
  • N-Methyl-D-glucamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.