missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-(isoquinolin-4-ylmethyl)amine, 97%, Thermo Scientific™

105.00 € - 272.00 €

Chemical Identifiers

CAS 157610-82-7
Molecular Formula C11H12N2
Molecular Weight (g/mol) 172.23
MDL Number MFCD09879899
InChI Key PISGLKXJHQKIFN-UHFFFAOYSA-N
Synonym n-methyl-isoquinolin-4-ylmethyl amine, isoquinolin-4-ylmethyl methyl amine, 4-isoquinolylmethyl methylamine, 4-isoquinolinemethanamine, n-methyl, n-methyl-iso quinolin-4-yl methyl amine
PubChem CID 24229453
IUPAC Name 1-isoquinolin-4-yl-N-methylmethanamine
SMILES CNCC1=C2C=CC=CC2=CN=C1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10553924
View Documents
Thermo Scientific Maybridge
CC04946CB
250 mg Amber Glass Bottle
105.00 €
250mg
Please to purchase this item. Need a web account? Register with us today!
10788045
View Documents
Thermo Scientific Maybridge
CC04946DA
1 g Amber Glass Bottle
272.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

157610-82-7
172.23
PISGLKXJHQKIFN-UHFFFAOYSA-N
24229453
CNCC1=C2C=CC=CC2=CN=C1
C11H12N2
MFCD09879899
n-methyl-isoquinolin-4-ylmethyl amine, isoquinolin-4-ylmethyl methyl amine, 4-isoquinolylmethyl methylamine, 4-isoquinolinemethanamine, n-methyl, n-methyl-iso quinolin-4-yl methyl amine
1-isoquinolin-4-yl-N-methylmethanamine

Specifications

157610-82-7
100.0
C11H12N2
n-methyl-isoquinolin-4-ylmethyl amine, isoquinolin-4-ylmethyl methyl amine, 4-isoquinolylmethyl methylamine, 4-isoquinolinemethanamine, n-methyl, n-methyl-iso quinolin-4-yl methyl amine
CNCC1=C2C=CC=CC2=CN=C1
172.23
172.23
N-methyl-(isoquinolin-4-ylmethyl)amine
96.5
97%
MFCD09879899
PISGLKXJHQKIFN-UHFFFAOYSA-N
1-isoquinolin-4-yl-N-methylmethanamine
24229453
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.