missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-methyl-N-(2-phenoxybenzyl)amine, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Spezifikation
Spezifikation
Assay Percent Range | 95% |
Quantity | 250 mg |
Formula Weight | 213.28 |
Percent Purity | 95% |
Chemical Name or Material | N-Methyl-N-(2-phenoxybenzyl)amine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'