missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-[(2-piperidin-1-ylpyridin-4-yl)methyl]amine, 97%, Thermo Scientific™

Product Code. 10517474
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 879896-58-9
Molecular Formula C12H19N3
Molecular Weight (g/mol) 205.305
MDL Number MFCD09025859
InChI Key CPEUGLUNQVAGDV-UHFFFAOYSA-N
Synonym n-methyl-n-2-piperidin-1-ylpyridin-4-yl methyl amine, methyl 2-piperidin-1-yl pyridin-4-yl methyl amine, 4-pyridinemethanamine,n-methyl-2-1-piperidinyl, n-methyl-1-2-piperidin-1-yl pyridin-4-yl methanamine, methyl 2-piperidyl 4-pyridyl methyl amine
PubChem CID 17851834
IUPAC Name N-methyl-1-(2-piperidin-1-ylpyridin-4-yl)methanamine
SMILES CNCC1=CC(=NC=C1)N2CCCCC2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 205.3
Percent Purity 97%
Chemical Name or Material N-Methyl-N-[(2-piperidin-1-ylpyridin-4-yl)methyl]amine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'