missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-[3-(1H-1,2,4-triazol-1-yl)benzyl]amine 97+%, Thermo Scientific™

Product Code. 10750824
Click to view available options
Quantity:
250 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 879896-43-2
Assay Percent Range 97+%
Molecular Formula C10H12N4
MDL Number MFCD08435864
Quantity 250 mg
Synonym n-methyl-n-3-1h-1,2,4-triazol-1-yl benzyl amine, methyl 3-1,2,4-triazol-1-yl phenyl methyl amine, benzenemethanamine,n-methyl-3-1h-1,2,4-triazol-1-yl, methyl 3-1,2,4-triazolyl phenyl methyl amine, n-methyl-n-3-1h-1,2,4-trazol-1-yl benzyl amne, methyl 3-1h-1,2,4-triazol-1-yl phenyl methyl amine, 1-3-1h-1,2,4-triazol-1-yl phenyl-n-methylmethanamine, n-methyl-1-3-1h-1,2,4-triazol-1-yl phenyl methanamine
InChI Key LLGQTEDUTKNYIP-UHFFFAOYSA-N
SMILES CNCC1=CC(=CC=C1)N2C=NC=N2
IUPAC Name N-methyl-1-[3-(1,2,4-triazol-1-yl)phenyl]methanamine
Molecular Weight (g/mol) 188.234
PubChem CID 18525742
Formula Weight 188.23
Chemical Name or Material n-methyl-n-[3-(1h-1, 2, 4-triazol-1-yl)benzyl]amine
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'