missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-[3-(trifluoromethyl)benzyl]amine, 97+%, Thermo Scientific™

Product Code. 10412674
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 90390-07-1
Molecular Formula C9H10F3N
Molecular Weight (g/mol) 189.181
InChI Key JFLPPELZYKHKQZ-UHFFFAOYSA-N
Synonym n-methyl-3-trifluoromethyl benzylamine, n-methyl-n-3-trifluoromethyl benzyl amine, n-methyl-1-3-trifluoromethyl phenyl methanamine, methyl 3-trifluoromethyl phenyl methyl amine, benzenemethanamine, n-methyl-3-trifluoromethyl, n-methyl 3-trifluoromethyl phenyl methanamine, n-methyl-1-3-trifluoromethyl phenyl methanamine hydrochloride, chembrdg-bb 4004318, methyl-3-trifluoromethyl-benzyl-amine
PubChem CID 485414
IUPAC Name N-methyl-1-[3-(trifluoromethyl)phenyl]methanamine
SMILES CNCC1=CC(=CC=C1)C(F)(F)F

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 189.18
Percent Purity 97%
Chemical Name or Material N-Methyl-N-[3-(trifluoromethyl)benzyl]amine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'