missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-((4-methyl-2-phenyl-1,3-thiazol-5-yl)methyl)amine, 97%, Thermo Scientific™

Product Code. 10661465
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 850375-02-9
Molecular Formula C12H14N2S
Molecular Weight (g/mol) 218.318
MDL Number MFCD07368511
InChI Key GFAGRBRYZWAUSV-UHFFFAOYSA-N
Synonym n-methyl-n-4-methyl-2-phenyl-1,3-thiazol-5-yl methyl amine, n-methyl-1-4-methyl-2-phenyl-1,3-thiazol-5-yl methanamine, methyl 4-methyl-2-phenyl-1,3-thiazol-5-yl methyl amine, 5-thiazolemethanamine,n,4-dimethyl-2-phenyl, n-methyl-1-4-methyl-2-phenylthiazol-5-yl methanamine, methyl 4-methyl-2-phenyl 1,3-thiazol-5-yl methyl amine, mmj
PubChem CID 4961253
IUPAC Name N-methyl-1-(4-methyl-2-phenyl-1,3-thiazol-5-yl)methanamine
SMILES CC1=C(SC(=N1)C2=CC=CC=C2)CNC

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 218.32
Percent Purity 97%
Chemical Name or Material N-methyl-N-[(4-methyl-2-phenyl-1, 3-thiazol-5-yl)methyl]amine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'