missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-[4-(pyridin-2-yloxy)benzyl]amine, 97%, Thermo Scientific™

Product Code. 10768655
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 892501-90-5
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C13H14N2O
MDL Number MFCD08690279
Quantity 1 g
Synonym n-methyl-n-4-pyridin-2-yloxy benzyl amine, methyl 4-pyridin-2-yloxy phenyl methyl amine, methyl 4-2-pyridyloxy phenyl methyl amine, benzenemethanamine,n-methyl-4-2-pyridinyloxy, n-methyl-1-4-pyridin-2-yloxyphenyl methanamine, n-methyl-1-4-2-pyridinyloxy phenyl methanamine, n-methyl-1-4-pyridin-2-yloxy phenyl methanamine, n-methyl-1-4-pyridin-2-yl oxy phenyl methanamine
InChI Key AFHNQZMRTDPSDR-UHFFFAOYSA-N
SMILES CNCC1=CC=C(OC2=CC=CC=N2)C=C1
IUPAC Name N-methyl-1-(4-pyridin-2-yloxyphenyl)methanamine
Molecular Weight (g/mol) 214.27
PubChem CID 18525833
Formula Weight 214.27
Percent Purity 97%
Chemical Name or Material N-methyl-N-[4-(pyridin-2-yloxy)benzyl]amine
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'