missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-[(5-morpholin-4-ylthien-2-yl)methyl]amine, 97%, Thermo Scientific™

61.83 € - 157.16 €

Chemical Identifiers

CAS 879896-62-5
Molecular Formula C10H16N2OS
Molecular Weight (g/mol) 212.311
MDL Number MFCD09025876
InChI Key LTBKEKXCWIEXPM-UHFFFAOYSA-N
Synonym n-methyl-n-5-morpholin-4-ylthien-2-yl methyl amine, methyl 5-morpholin-4-yl thiophen-2-yl methyl amine, methyl 5-morpholin-4-yl 2-thienyl methyl amine, 2-thiophenemethanamine,n-methyl-5-4-morpholinyl, n-methyl-1-5-morpholin-4-yl thiophen-2-yl methanamine
PubChem CID 18525863
IUPAC Name N-methyl-1-(5-morpholin-4-ylthiophen-2-yl)methanamine
SMILES CNCC1=CC=C(S1)N2CCOCC2
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
11356785
View Documents Promotion Details
Thermo Scientific Maybridge
CC53446CB
250 mg Amber Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10012414
View Documents Promotion Details
Thermo Scientific Maybridge
CC53446DA
1 g Amber Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

879896-62-5
212.311
LTBKEKXCWIEXPM-UHFFFAOYSA-N
18525863
CNCC1=CC=C(S1)N2CCOCC2
C10H16N2OS
MFCD09025876
n-methyl-n-5-morpholin-4-ylthien-2-yl methyl amine, methyl 5-morpholin-4-yl thiophen-2-yl methyl amine, methyl 5-morpholin-4-yl 2-thienyl methyl amine, 2-thiophenemethanamine,n-methyl-5-4-morpholinyl, n-methyl-1-5-morpholin-4-yl thiophen-2-yl methanamine
N-methyl-1-(5-morpholin-4-ylthiophen-2-yl)methanamine

Specifications

879896-62-5
100.0
C10H16N2OS
n-methyl-n-5-morpholin-4-ylthien-2-yl methyl amine, methyl 5-morpholin-4-yl thiophen-2-yl methyl amine, methyl 5-morpholin-4-yl 2-thienyl methyl amine, 2-thiophenemethanamine,n-methyl-5-4-morpholinyl, n-methyl-1-5-morpholin-4-yl thiophen-2-yl methanamine
CNCC1=CC=C(S1)N2CCOCC2
212.311
212.32
N-methyl-N-[(5-morpholin-4-ylthien-2-yl)methyl]amine
96.5
97%
MFCD09025876
LTBKEKXCWIEXPM-UHFFFAOYSA-N
N-methyl-1-(5-morpholin-4-ylthiophen-2-yl)methanamine
18525863
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications