missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methylaniline, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2500 g
Description
N-Methylaniline is used in the preparation of N,N'-dimethylsulfinyldianilin using triethylamine and thionyl chloride as reagents. It is also acts as a latent and coupling solvent. Further, it serves as an intermediate for dyes and agrochemicals. In addition to this, it is utilized as an antiknock agent, which plays an essential role to increase the octane number.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Methylaniline is used in the preparation of N,N′-dimethylsulfinyldianilin using triethylamine and thionyl chloride as reagents. It is also acts as a latent and coupling solvent. Further, it serves as an intermediate for dyes and agrochemicals. In addition to this, it is utilized as an antiknock agent, which plays an essential role to increase the octane number.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents, acids, acid anhydrides, acid chlorides and carbon dioxide.
N-Methylaniline is used in the preparation of N,N′-dimethylsulfinyldianilin using triethylamine and thionyl chloride as reagents. It is also acts as a latent and coupling solvent. Further, it serves as an intermediate for dyes and agrochemicals. In addition to this, it is utilized as an antiknock agent, which plays an essential role to increase the octane number.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents, acids, acid anhydrides, acid chlorides and carbon dioxide.
Specifications
Melting Point | -57°C |
Density | 0.989 |
Boiling Point | 194°C to 197°C |
Flash Point | 86°C (186°F) |
Refractive Index | 1.571 |
Quantity | 100 g |
UN Number | UN2294 |
Beilstein | 741982 |
Sensitivity | Air Sensitive |
Merck Index | 14,6019 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'