Learn More
N-Methylaniline, 99%, Thermo Scientific Chemicals
55.48 € - 194.97 €
Chemical Identifiers
CAS | 100-61-8 |
---|---|
Molecular Formula | C7H9N |
Molecular Weight (g/mol) | 107.16 |
MDL Number | MFCD00008283 |
InChI Key | AFBPFSWMIHJQDM-UHFFFAOYSA-N |
Synonym | methylaniline, monomethylaniline, n-methyl aniline, benzenamine, n-methyl, methylphenylamine, n-methylbenzenamine, n-monomethylaniline, n-methylaminobenzene, methylamino benzene, anilinomethane |
PubChem CID | 7515 |
ChEBI | CHEBI:15733 |
IUPAC Name | N-methylaniline |
SMILES | CNC1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10408670
|
Thermo Scientific Acros
126275000 |
500 g | Glass bottle |
55.48 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10703891
|
Thermo Scientific Acros
126270010 |
1 kg | Glass bottle |
87.23 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10023223
|
Thermo Scientific Acros
126270025 |
2.5 kg | Glass bottle |
194.97 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
100-61-8 | |
107.16 | |
AFBPFSWMIHJQDM-UHFFFAOYSA-N | |
7515 | |
N-methylaniline |
C7H9N | |
MFCD00008283 | |
methylaniline, monomethylaniline, n-methyl aniline, benzenamine, n-methyl, methylphenylamine, n-methylbenzenamine, n-monomethylaniline, n-methylaminobenzene, methylamino benzene, anilinomethane | |
CHEBI:15733 | |
CNC1=CC=CC=C1 |
Specifications
100-61-8 | |
100.0 | |
Brown to Yellow | |
195.0°C | |
Authentic | |
C7H9N | |
1.5694 to 1.5714 | |
12, 135 | |
15, 6093 | |
methylaniline, monomethylaniline, n-methyl aniline, benzenamine, n-methyl, methylphenylamine, n-methylbenzenamine, n-monomethylaniline, n-methylaminobenzene, methylamino benzene, anilinomethane | |
AFBPFSWMIHJQDM-UHFFFAOYSA-N | |
N-methylaniline | |
7515 | |
107.15 | |
Liquid |
98.5 | |
-57.0°C | |
0.9870g/mL | |
86°C | |
98.5% min. (GC) | |
C6H5NHCH3 | |
MFCD00008283 | |
14,207 | |
0.987 | |
Solubility in water: 30g/l. Other solubilities: soluble in alcohol and ether,soluble in acetone,diethyl ether,ethanol and,chloroform | |
CNC1=CC=CC=C1 | |
107.16 | |
CHEBI:15733 | |
99% | |
N-Methylaniline |
Safety and Handling
GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Toxic if inhaled.
Toxic in contact with skin.
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 202-870-9
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.