Learn More
N-Methylhydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals
Description
N-Methylhydroxylamine hydrochloride is used as an inorganic catalyst used in the transamidation of primary amides with amines. It is employed in constructing hydroxamates, important functional groups for the complexation of iron.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
N-Methylhydroxylamine hydrochloride is used as an inorganic catalyst used in the transamidation of primary amides with amines. It is employed in constructing hydroxamates, important functional groups for the complexation of iron.
Solubility
Soluble in water.
Notes
Hygroscopic. Store under dry inert gas. Protect from humidity and air.
Specifications
Specifications
Melting Point | 80°C to 86°C |
Quantity | 25 g |
Beilstein | 3541409 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water. |
Formula Weight | 83.52 |
Percent Purity | 98% |
Chemical Name or Material | N-Methylhydroxylamine hydrochloride |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.