missing translation for 'onlineSavingsMsg'
Learn More

N-Methylmorpholine, 99%, Thermo Scientific Chemicals

25.53 € - 223.00 €

Chemical Identifiers

CAS 109-02-4
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.15
MDL Number MFCD00006175
InChI Key SJRJJKPEHAURKC-UHFFFAOYSA-N
Synonym n-methylmorpholine, morpholine, 4-methyl, methylmorpholine, 1-methylmorpholine, morpholine, n-methyl, 4-methylmorfolin, 4-methylmorpholin, n-methyl morpholine, 4-methylmorfolin czech, n-methylmorpholin
PubChem CID 7972
IUPAC Name 4-methylmorpholine
SMILES CN1CCOCC1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10626712
View Documents
Thermo Scientific Acros
127151000
100 g Glass Bottle
25.53 €
100g
Please to purchase this item. Need a web account? Register with us today!
10626902
View Documents
Thermo Scientific Acros
127155000
500 g Glass Bottle
73.57 €
500g
Please to purchase this item. Need a web account? Register with us today!
11966801
View Documents
Thermo Scientific Acros
127150025
2.5 kg Plastic drum
223.00 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

109-02-4
101.15
SJRJJKPEHAURKC-UHFFFAOYSA-N
7972
CN1CCOCC1
C5H11NO
MFCD00006175
n-methylmorpholine, morpholine, 4-methyl, methylmorpholine, 1-methylmorpholine, morpholine, n-methyl, 4-methylmorfolin, 4-methylmorpholin, n-methyl morpholine, 4-methylmorfolin czech, n-methylmorpholin
4-methylmorpholine

Specifications

109-02-4
100.0
0.9200g/mL
14°C
98.5% min. (GC)
1.4339 to 1.4359
27,6
15,6362
n-methylmorpholine, morpholine, 4-methyl, methylmorpholine, 1-methylmorpholine, morpholine, n-methyl, 4-methylmorfolin, 4-methylmorpholin, n-methyl morpholine, 4-methylmorfolin czech, n-methylmorpholin
SJRJJKPEHAURKC-UHFFFAOYSA-N
4-methylmorpholine
7972
101.15
Liquid
98.5
-65.0°C
115.0°C to 116.0°C (750.0 mmHg)
Authentic
C5H11NO
MFCD00006175
01,690; 02,278
0.92
Solubility in water: >500g/L (20°C). Other solubilities: miscible with alcohol,ether and benzene
CN1CCOCC1
101.15
0.91 mPa.s (20°C)
99%
N-Methylmorpholine, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Highly flammable liquid and vapor.

GHS P Statement
Ground/Bond container and receiving equipment.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 203-640-0

missing translation for 'rtecsNumber' : QE5775000

missing translation for 'tsca' : TSCA

Product Identifier
  • N-Methylmorpholine

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.