missing translation for 'onlineSavingsMsg'
Learn More

N-Methylpiperidine, 99%, Thermo Scientific Chemicals

Product Code. 10582521
Click to view available options
Quantity:
10 mL
250 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-67-5
Molecular Formula C6H14ClN
Molecular Weight (g/mol) 135.64
MDL Number MFCD00006491
InChI Key QDUXDCXILAPLAG-UHFFFAOYSA-N
Synonym n-methylpiperidine, piperidine, 1-methyl, methylpiperidine, n-methyl piperidine, 1-methyl-piperidine, n-methylpiperidin, methylpiperdine, 1-methylpiperadine, n-methyl-piperidine, 1-methyl piperidine
PubChem CID 12291
IUPAC Name 1-methylpiperidine
SMILES [H+].[Cl-].CN1CCCCC1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -50.0°C
Color Colorless
Density 0.8200g/mL
Boiling Point 105.0°C to 108.0°C
Flash Point 3°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4368 to 1.4388
Quantity 250 mL
Beilstein 20,16
Specific Gravity 0.82
Solubility Information Solubility in water: miscible. Other solubilities: soluble in methanol, ethanol, acetone, toluene and
Viscosity 5 mPa.s (20°C)
Formula Weight 99.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N-Methylpiperidine, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N-Methylpiperidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.