missing translation for 'onlineSavingsMsg'
Learn More

N-Methylsulfanilic acid, 98+%, Thermo Scientific™

34.35 € - 119.69 €

Chemical Identifiers

CAS 24447-99-2
Molecular Formula C7H9NO3S
Molecular Weight (g/mol) 187.213
MDL Number MFCD00014732
InChI Key QRAXZXPSAGQUNP-UHFFFAOYSA-N
Synonym 4-methylamino benzenesulfonic acid, 4-methylamino benzenesulphonic acid, n-methylsulphanilic acid, n-methylsulfanilic acid, benzenesulfonic acid,4-methylamino, n-methylaniline-p-sulfonic acid, maybridge1_004198, n-methylsulfanilic acid, 98+%
PubChem CID 90505
IUPAC Name 4-(methylamino)benzenesulfonic acid
SMILES CNC1=CC=C(C=C1)S(=O)(=O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11384414
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L01016.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11394414
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L01016.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

24447-99-2
187.213
QRAXZXPSAGQUNP-UHFFFAOYSA-N
90505
CNC1=CC=C(C=C1)S(=O)(=O)O
C7H9NO3S
MFCD00014732
4-methylamino benzenesulfonic acid, 4-methylamino benzenesulphonic acid, n-methylsulphanilic acid, n-methylsulfanilic acid, benzenesulfonic acid,4-methylamino, n-methylaniline-p-sulfonic acid, maybridge1_004198, n-methylsulfanilic acid, 98+%
4-(methylamino)benzenesulfonic acid

Specifications

24447-99-2
C7H9NO3S
2094129
QRAXZXPSAGQUNP-UHFFFAOYSA-N
4-(methylamino)benzenesulfonic acid
90505
≥98%
∽260°C (decomposition)
MFCD00014732
4-methylamino benzenesulfonic acid, 4-methylamino benzenesulphonic acid, n-methylsulphanilic acid, n-methylsulfanilic acid, benzenesulfonic acid,4-methylamino, n-methylaniline-p-sulfonic acid, maybridge1_004198, n-methylsulfanilic acid, 98+%
CNC1=CC=C(C=C1)S(=O)(=O)O
187.213
187.21
N-Methylsulfanilic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 246-256-9

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications