missing translation for 'onlineSavingsMsg'
Learn More

N,N-Bis(2-hydroxyethyl)ethylenediamine, Thermo Scientific Chemicals

Product Code. 11348347
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is employed as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3197-06-6
Molecular Formula C6H16N2O2
Molecular Weight (g/mol) 148.21
MDL Number MFCD00014822
InChI Key CYOIAXUAIXVWMU-UHFFFAOYSA-N

Specifications

Density 1.069 g/mL
Boiling Point 174°C to 178°C (3 to 4 mmHg)
Odor Amine-like
Quantity 1 g
UN Number UN2735
Synonym n,n-bis 2-hydroxyethyl ethylenediamine, 2,2'-2-aminoethyl azanediyl diethanol, ethanol, 2,2'-2-aminoethyl imino bis, 2,2'-2-aminoethyl imino bisethanol, 2-2-aminoethyl 2-hydroxyethyl amino ethanol, 2-2-aminoethyl 2-hydroxyethyl amino ethan-1-ol, aminoethyldiethanolamine, aminoethylethanolamine solution, 2,2'-2-aminoethyl imino diethanol, acmc-20a9el
Solubility Information Soluble in water.
SMILES C(CN(CCO)CCO)N
IUPAC Name 2-[2-aminoethyl(2-hydroxyethyl)amino]ethanol
PubChem CID 98535
Formula Weight 148.21
Physical Form Liquid
Chemical Name or Material N,N-Bis(2-hydroxyethyl)ethylenediamine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.