missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(benzyloxycarbonyl)-1H-pyrazole-1-carboxamidine, 98+%, Thermo Scientific Chemicals

Product Code. 15450756
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

N,N'-Bis(benzyloxycarbonyl)-1H-pyrazole-1-carboxamidine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Bis(benzyloxycarbonyl)-1H-pyrazole-1-carboxamidine is used as pharmaceutical intermediate.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 152120-55-3
Molecular Formula C20H18N4O4
Molecular Weight (g/mol) 378.388
MDL Number MFCD02683516
InChI Key NRBUVVTTYMTSKM-UHFFFAOYSA-N
Synonym n,n'-bis-z-1-guanylpyrazole, n,n'-bis benzyloxycarbonyl-1h-pyrazole-1-carboxamidine, n,n-bisbenzyloxycarbonyl-1-guanylpyrazole, benzyl benzyloxy carbonyl amino 1h-pyrazol-1-yl methylene carbamate, pyrazole z 2, pyrazol z 2, n,n'-bs-z-1-guanylpyrazole, n,n;-bis-z-1-guanylpyrazole, benzyl n-1z-benzyloxy carbonyl imino pyrazol-1-yl methyl carbamate, n,n'-di-cbz-1h-pyrazole-1-carbo
PubChem CID 6371643
IUPAC Name benzyl (NZ)-N-[phenylmethoxycarbonylamino(pyrazol-1-yl)methylidene]carbamate
SMILES C1=CC=C(C=C1)COC(=O)NC(=NC(=O)OCC2=CC=CC=C2)N3C=CC=N3

Specifications

Melting Point 87°C to 91°C
Quantity 1 g
Formula Weight 378.38
Percent Purity ≥98%
Chemical Name or Material N,N'-Bis(benzyloxycarbonyl)-1H-pyrazole-1-carboxamidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'