missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Bis(Carboxymethyl)-Dl-Alanine Trisodium Salt, ≥90.0%, Honeywell™ Fluka™
Click to view available options
Quantity:
100 g
Packaging:
Poly bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
CAS | 164462-16-2 |
Packaging | Poly bottle |
Molecular Formula | C7H11NNa3O6 |
MDL Number | MFCD08276427 |
Quantity | 100 g |
UN Number | NONH for all modes of transport |
Synonym | n,n-bis carboxylatomethyl alanine trisodium salt |
InChI Key | SQKOOOSPGWXVRN-UHFFFAOYSA-N |
SMILES | CC(C(=O)O)N(CC(=O)O)CC(=O)O.[Na].[Na].[Na] |
IUPAC Name | 2-[bis(carboxymethyl)amino]propanoic acid;sodium |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'