missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(salicylidene)ethylenediaminecobalt(II), 96%, Thermo Scientific Chemicals

Product Code. 11344858
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Salcomine catalyzes the oxidation of 2,6-disubstituted phenols by dioxygen. Experimental inhibitor of human cytμloviris proteinase activity. Metal complex dyes used as redox catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14167-18-1
Molecular Formula C16H14CoN2O2
Molecular Weight (g/mol) 325.23
MDL Number MFCD00000009
InChI Key NPAQSKHBTMUERN-UHFFFAOYSA-L
Synonym Salcomine
PubChem CID 57448789
SMILES [Co++].[O-]C1=CC=CC=C1C=NCCN=CC1=CC=CC=C1[O-]

Specifications

Color Black
Odor Odorless
Linear Formula [-CH2N=CH(C6H4O-)]2Co
Quantity 5 g
Solubility Information Insoluble in water. Soluble in benzene,chloroform,and pyridine.
IUPAC Name cobalt;(6Z)-6-[[2-[[(E)-(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino]ethylamino]methylidene]cyclohexa-2,4-dien-1-one
Formula Weight 325.23
Percent Purity 96%
Physical Form Powder
Chemical Name or Material N,N'-Bis (salicylidene)ethylenediaminecobalt (II)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'