missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(salicylidene)ethylenediaminecobalt(II), 96%, Thermo Scientific Chemicals

Product Code. 11354858
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Salcomine catalyzes the oxidation of 2,6-disubstituted phenols by dioxygen. Experimental inhibitor of human cytμloviris proteinase activity. Metal complex dyes used as redox catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14167-18-1
Molecular Formula C16H14CoN2O2
Molecular Weight (g/mol) 325.23
MDL Number MFCD00000009
InChI Key NPAQSKHBTMUERN-UHFFFAOYSA-L
Synonym Salcomine
PubChem CID 57448789
SMILES [Co++].[O-]C1=CC=CC=C1C=NCCN=CC1=CC=CC=C1[O-]

Specifications

Color Black
Odor Odorless
Linear Formula [-CH2N=CH(C6H4O-)]2Co
Quantity 100 g
Solubility Information Insoluble in water. Soluble in benzene,chloroform,and pyridine.
IUPAC Name cobalt;(6Z)-6-[[2-[[(E)-(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino]ethylamino]methylidene]cyclohexa-2,4-dien-1-one
Formula Weight 325.23
Percent Purity 96%
Physical Form Powder
Chemical Name or Material N,N'-Bis (salicylidene)ethylenediaminecobalt (II)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.