missing translation for 'onlineSavingsMsg'
Learn More

N-(n-Butyl)aniline, 99%, Thermo Scientific Chemicals

50.00 € - 162.00 €

Chemical Identifiers

CAS 1126-78-9
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.237
MDL Number MFCD00035798
InChI Key VSHTWPWTCXQLQN-UHFFFAOYSA-N
Synonym benzenamine, n-butyl, aniline, n-butyl, 4-phenylamino butane, n-butylbenzenamine, n-n-butyl aniline, butylaniline, n-phenyl-n-butylamine, butylphenylamine, unii-r2znj7l2um, benzamine, n-butyl
PubChem CID 14310
IUPAC Name N-butylaniline
SMILES CCCCNC1=CC=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11411217
View Documents
Thermo Scientific Alfa Aesar
B24180.14
25 g
50.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
11421217
View Documents
Thermo Scientific Alfa Aesar
B24180.22
100 g
162.00 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1126-78-9
149.237
VSHTWPWTCXQLQN-UHFFFAOYSA-N
14310
CCCCNC1=CC=CC=C1
C10H15N
MFCD00035798
benzenamine, n-butyl, aniline, n-butyl, 4-phenylamino butane, n-butylbenzenamine, n-n-butyl aniline, butylaniline, n-phenyl-n-butylamine, butylphenylamine, unii-r2znj7l2um, benzamine, n-butyl
N-butylaniline

Specifications

1126-78-9
240°C to 242°C
C10H15N
MFCD00035798
benzenamine, n-butyl, aniline, n-butyl, 4-phenylamino butane, n-butylbenzenamine, n-n-butyl aniline, butylaniline, n-phenyl-n-butylamine, butylphenylamine, unii-r2znj7l2um, benzamine, n-butyl
CCCCNC1=CC=CC=C1
149.237
149.23
N-(n-Butyl)aniline
0.932
105°C (222°F)
1.532
UN2738
VSHTWPWTCXQLQN-UHFFFAOYSA-N
N-butylaniline
14310
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H332

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: N-BUTYLANILINE

missing translation for 'einecsNumber' : 214-425-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.