missing translation for 'onlineSavingsMsg'
Learn More

N-n-Butyldiethanolamine, 98%, Thermo Scientific Chemicals

Product Code. 11352367
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-79-4
Molecular Formula C8H19NO2
Molecular Weight (g/mol) 161.245
MDL Number MFCD00002856
InChI Key GVNHOISKXMSMPX-UHFFFAOYSA-N
Synonym n-butyldiethanolamine, butyldiethanolamine, ethanol, 2,2'-butylimino bis, bide, 2,2'-butylimino diethanol, n,n-bis 2-hydroxyethyl butylamine, butylbis 2-hydroxyethyl amine, n-butyl-2,2'-iminodiethanol, 2,2-butylimino diethanol, n-butyl-n,n-bis hydroxyethyl amine
PubChem CID 7620
IUPAC Name 2-[butyl(2-hydroxyethyl)amino]ethanol
SMILES CCCCN(CCO)CCO

Specifications

Melting Point -70°C
Density 0.968
Boiling Point 268°C to 272°C
Flash Point 143°C (289°F)
Refractive Index 1.464
Quantity 500 g
UN Number UN2735
Beilstein 1739642
Sensitivity Hygroscopic
Formula Weight 161.25
Percent Purity 98%
Chemical Name or Material N-n-Butyldiethanolamine
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'