missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N'-Di(1-naphthyl)-N,N'-diphenylbenzidine, 98%, Thermo Scientific Chemicals
49.75 € - 368.00 €
Chemical Identifiers
CAS | 123847-85-8 |
---|---|
Molecular Formula | C44H32N2 |
Molecular Weight (g/mol) | 588.754 |
MDL Number | MFCD03093246 |
InChI Key | IBHBKWKFFTZAHE-UHFFFAOYSA-N |
Synonym | n4,n4'-di naphthalen-1-yl-n4,n4'-diphenyl-1,1'-biphenyl-4,4'-diamine, alpha-npb, n,n'-bis naphthalen-1-yl-n,n'-bis phenyl benzidine, 4,4'-bis n-1-naphthyl-n-phenylamino biphenyl, n,n'-di 1-naphthyl-n,n'-diphenylbenzidine, n,n'-bis-1-naphthalenyl-n,n'-bis-phenyl-1,1'-biphenyl-4,4'-diamine, 1,1'-biphenyl-4,4'-diamine, n,n'-di-1-naphthalenyl-n,n'-diphenyl, n,n'-di-1-naphthyl-n,n'-diphenylbenzidine |
PubChem CID | 5069127 |
IUPAC Name | N-[4-[4-(N-naphthalen-1-ylanilino)phenyl]phenyl]-N-phenylnaphthalen-1-amine |
SMILES | C1=CC=C(C=C1)N(C2=CC=C(C=C2)C3=CC=C(C=C3)N(C4=CC=CC=C4)C5=CC=CC6=CC=CC=C65)C7=CC=CC8=CC=CC=C87 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15478956
|
Thermo Scientific Alfa Aesar
H64502.MD |
250 mg |
49.75 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15411749
|
Thermo Scientific Alfa Aesar
H64502.03 |
1 g |
103.52 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15421749
|
Thermo Scientific Alfa Aesar
H64502.06 |
5 g |
368.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
123847-85-8 | |
588.754 | |
IBHBKWKFFTZAHE-UHFFFAOYSA-N | |
5069127 | |
C1=CC=C(C=C1)N(C2=CC=C(C=C2)C3=CC=C(C=C3)N(C4=CC=CC=C4)C5=CC=CC6=CC=CC=C65)C7=CC=CC8=CC=CC=C87 |
C44H32N2 | |
MFCD03093246 | |
n4,n4'-di naphthalen-1-yl-n4,n4'-diphenyl-1,1'-biphenyl-4,4'-diamine, alpha-npb, n,n'-bis naphthalen-1-yl-n,n'-bis phenyl benzidine, 4,4'-bis n-1-naphthyl-n-phenylamino biphenyl, n,n'-di 1-naphthyl-n,n'-diphenylbenzidine, n,n'-bis-1-naphthalenyl-n,n'-bis-phenyl-1,1'-biphenyl-4,4'-diamine, 1,1'-biphenyl-4,4'-diamine, n,n'-di-1-naphthalenyl-n,n'-diphenyl, n,n'-di-1-naphthyl-n,n'-diphenylbenzidine | |
N-[4-[4-(N-naphthalen-1-ylanilino)phenyl]phenyl]-N-phenylnaphthalen-1-amine |
Specifications
123847-85-8 | |
Yellow | |
MFCD03093246 | |
n4,n4'-di naphthalen-1-yl-n4,n4'-diphenyl-1,1'-biphenyl-4,4'-diamine, alpha-npb, n,n'-bis naphthalen-1-yl-n,n'-bis phenyl benzidine, 4,4'-bis n-1-naphthyl-n-phenylamino biphenyl, n,n'-di 1-naphthyl-n,n'-diphenylbenzidine, n,n'-bis-1-naphthalenyl-n,n'-bis-phenyl-1,1'-biphenyl-4,4'-diamine, 1,1'-biphenyl-4,4'-diamine, n,n'-di-1-naphthalenyl-n,n'-diphenyl, n,n'-di-1-naphthyl-n,n'-diphenylbenzidine | |
C1=CC=C(C=C1)N(C2=CC=C(C=C2)C3=CC=C(C=C3)N(C4=CC=CC=C4)C5=CC=CC6=CC=CC=C65)C7=CC=CC8=CC=CC=C87 | |
588.754 | |
588.75 | |
Powder |
279°C to 283°C | |
C44H32N2 | |
8022378 | |
IBHBKWKFFTZAHE-UHFFFAOYSA-N | |
N-[4-[4-(N-naphthalen-1-ylanilino)phenyl]phenyl]-N-phenylnaphthalen-1-amine | |
5069127 | |
98% | |
N,N'-Di(1-naphthyl)-N,N'-diphenylbenzidine |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only