Learn More
N,N'-Di-tert-butylethylenediamine, 98%, Thermo Scientific Chemicals
51.56 € - 166.83 €
Chemical Identifiers
CAS | 4062-60-6 |
---|---|
Molecular Formula | C10H24N2 |
Molecular Weight (g/mol) | 172.316 |
MDL Number | MFCD00014996 |
InChI Key | KGHYGBGIWLNFAV-UHFFFAOYSA-N |
Synonym | n,n'-di-tert-butylethylenediamine, 1,2-bis tert-butylamino ethane, n,n'-di-t-butylethylenediamine, n1,n2-di-tert-butylethane-1,2-diamine, unii-6wai8u5v0w, n,n'-bis tert-butyl ethylenediamine, 1,2-ethanediamine, n,n'-bis 1,1-dimethylethyl, n,n'-di-tert-butyl-ethylenediamine, 6wai8u5v0w, n,n'-bis 1,1-dimethylethyl-1,2-ethanediamine |
PubChem CID | 77680 |
IUPAC Name | N,N'-ditert-butylethane-1,2-diamine |
SMILES | CC(C)(C)NCCNC(C)(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11301787
|
Thermo Scientific Alfa Aesar
L04972.06 |
5 g |
51.56 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11311787
|
Thermo Scientific Alfa Aesar
L04972.14 |
25 g |
166.83 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
N,N'-Di-tert-butylethylenediamine is used as a primary and secondary intermediate. It is also used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN,N′-Di-tert-butylethylenediamine is used as a primary and secondary intermediate. It is also used as a pharmaceutical intermediate.
Solubility
Soluble (33 g/L) (25°C),
Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, strong acids.
Chemical Identifiers
4062-60-6 | |
172.316 | |
KGHYGBGIWLNFAV-UHFFFAOYSA-N | |
77680 | |
CC(C)(C)NCCNC(C)(C)C |
C10H24N2 | |
MFCD00014996 | |
n,n'-di-tert-butylethylenediamine, 1,2-bis tert-butylamino ethane, n,n'-di-t-butylethylenediamine, n1,n2-di-tert-butylethane-1,2-diamine, unii-6wai8u5v0w, n,n'-bis tert-butyl ethylenediamine, 1,2-ethanediamine, n,n'-bis 1,1-dimethylethyl, n,n'-di-tert-butyl-ethylenediamine, 6wai8u5v0w, n,n'-bis 1,1-dimethylethyl-1,2-ethanediamine | |
N,N'-ditert-butylethane-1,2-diamine |
Specifications
4062-60-6 | |
62°C (143°F) | |
(CH3)3CNHCH2CH2NHC(CH3)3 | |
UN2735 | |
n,n'-di-tert-butylethylenediamine, 1,2-bis tert-butylamino ethane, n,n'-di-t-butylethylenediamine, n1,n2-di-tert-butylethane-1,2-diamine, unii-6wai8u5v0w, n,n'-bis tert-butyl ethylenediamine, 1,2-ethanediamine, n,n'-bis 1,1-dimethylethyl, n,n'-di-tert-butyl-ethylenediamine, 6wai8u5v0w, n,n'-bis 1,1-dimethylethyl-1,2-ethanediamine | |
CC(C)(C)NCCNC(C)(C)C | |
172.316 | |
172.32 | |
N,N'-Di-tert-butylethylenediamine |
132°C (100 mmHg) | |
C10H24N2 | |
MFCD00014996 | |
1738432 | |
KGHYGBGIWLNFAV-UHFFFAOYSA-N | |
N,N'-ditert-butylethane-1,2-diamine | |
77680 | |
98% |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: POLYAMINES, LIQUID, CORROSIVE, N.O.S.
missing translation for 'rtecsNumber' : KH8587137
missing translation for 'tsca' : TSCA
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.