missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dicyclohexylmethylamine, 97%

Product Code. 10432770 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
This item is not returnable. View return policy

Product Code. 10432770

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7560-83-0 | C13H25N | 195.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7560-83-0
Molecular Formula C13H25N
Molecular Weight (g/mol) 195.35
MDL Number MFCD00014289
InChI Key GSCCALZHGUWNJW-UHFFFAOYSA-N
Synonym n,n-dicyclohexylmethylamine, n-methyldicyclohexylamine, cyclohexanamine, n-cyclohexyl-n-methyl, dicyclohexylamine, n-methyl, n-methyl dicyclohexylamine, unii-ri95g6j8ff, n-cyclohexyl-n-methylcyclohexylamine, cy2nme, methyldicyclohexylamine, nn-dicyclohexylmethylamine
PubChem CID 24210
IUPAC Name N-cyclohexyl-N-methylcyclohexanamine
SMILES CN(C1CCCCC1)C2CCCCC2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Density 0.9210g/mL
Boiling Point 265.0°C
Flash Point 110°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 kg
Specific Gravity 0.921
Formula Weight 195.35
Percent Purity 97%
Chemical Name or Material N, N-Dicyclohexylmethylamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N,N-Dicyclohexylmethylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.