missing translation for 'onlineSavingsMsg'
Learn More
N,N-Diethyl-1,3-propanediamine, 99%, Thermo Scientific Chemicals Product Code.: 11373297

N,N-Diethyl-1,3-propanediamine, 99%, Thermo Scientific Chemicals

Product Code. 11373297
500 mL
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy

Product Code. 11373297

Brand: Thermo Scientific Alfa Aesar L13888.AP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

3-Diethylaminopropylamine is used as a solvent, extraction agents and organic synthesis intermediates, dyes, pigments, surface active agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Diethylaminopropylamine is used as a solvent, extraction agents and organic synthesis intermediates, dyes, pigments, surface active agent.

Solubility
Fully miscible in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-78-9
Molecular Formula C7H18N2
Molecular Weight (g/mol) 130.24
MDL Number MFCD00008218
InChI Key QOHMWDJIBGVPIF-UHFFFAOYSA-N
Synonym 3-diethylaminopropylamine, 3-diethylamino propylamine, n,n-diethyl-1,3-propanediamine, 3-aminopropyl diethylamine, n1,n1-diethylpropane-1,3-diamine, 1,3-propanediamine, n,n-diethyl, diethylaminotrimethylenamine, n,n-diethyl-1,3-diaminopropane, 1-diethylamino propylamine-3, n-3-diethylaminopropyl amineShow MoreShow Less
PubChem CID 61011
IUPAC Name N',N'-diethylpropane-1,3-diamine
SMILES CCN(CC)CCCN

Specifications

Melting Point -60°C
Density 0.826
Boiling Point 168°C to 171°C
Flash Point 51°C (124°F)
Linear Formula (CH3CH2)2N(CH2)3NH2
Refractive Index 1.442
Quantity 500 mL
UN Number UN2684
Beilstein 741879
Sensitivity Air Sensitive
Solubility Information Fully miscible in water.
Formula Weight 130.23
Percent Purity 99%
Chemical Name or Material N,N-Diethyl-1,3-propanediamine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0435679 Certificate TypeCertificate of Analysis Date25/11/2022 Catalog NumberL13888.AP,L13888.AE
Lot Number10223981 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL13888.AP,L13888.AE
Lot NumberGBFA000248 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL13888.AP,L13888.AE
Lot NumberL21R014 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL13888.AP,L13888.AE
Lot Number10130142 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL13888.AP,L13888.AE

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N,N-Diethyl-1,3-propanediamine, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.