Learn More
N,N'-Diethylethylenediamine, 95%, Thermo Scientific Chemicals
34.07 € - 117.59 €
Chemical Identifiers
CAS | 111-74-0 |
---|---|
Molecular Formula | C6H16N2 |
Molecular Weight (g/mol) | 116.21 |
MDL Number | MFCD00009033 |
InChI Key | CJKRXEBLWJVYJD-UHFFFAOYSA-N |
Synonym | n,n'-diethylethylenediamine, 1,2-ethanediamine, n,n'-diethyl, 1,2-bis ethylamino ethane, 3,6-diazaoctane, 1,2-ethanediamine, n1,n2-diethyl, n,n'-ethylenediethyldiamine, ethyl 2-ethylamino ethyl amine, ethylenediamine, n,n'-diethyl, n,n'-diethyl-1,2-diaminoethane, dihydrobromide |
PubChem CID | 67105 |
ChEBI | CHEBI:182290 |
IUPAC Name | N,N'-diethylethane-1,2-diamine |
SMILES | CCNCCNCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15274644
|
Thermo Scientific Acros
155520050 |
5 g | Glass bottle |
34.07 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15284644
|
Thermo Scientific Acros
155520250 |
25 g | Glass bottle |
117.59 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
111-74-0 | |
116.21 | |
CJKRXEBLWJVYJD-UHFFFAOYSA-N | |
67105 | |
N,N'-diethylethane-1,2-diamine |
C6H16N2 | |
MFCD00009033 | |
n,n'-diethylethylenediamine, 1,2-ethanediamine, n,n'-diethyl, 1,2-bis ethylamino ethane, 3,6-diazaoctane, 1,2-ethanediamine, n1,n2-diethyl, n,n'-ethylenediethyldiamine, ethyl 2-ethylamino ethyl amine, ethylenediamine, n,n'-diethyl, n,n'-diethyl-1,2-diaminoethane, dihydrobromide | |
CHEBI:182290 | |
CCNCCNCC |
Specifications
111-74-0 | |
152.0°C to 154.0°C | |
94% min. (GC) | |
C2H5NHCH2CH2NHC2H5 | |
MFCD00009033 | |
0.811 | |
Solubility in water: miscible. | |
CCNCCNCC | |
116.21 | |
CHEBI:182290 | |
95% |
0.8110g/mL | |
33°C | |
C6H16N2 | |
1.4326 | |
04,251 | |
n,n'-diethylethylenediamine, 1,2-ethanediamine, n,n'-diethyl, 1,2-bis ethylamino ethane, 3,6-diazaoctane, 1,2-ethanediamine, n1,n2-diethyl, n,n'-ethylenediethyldiamine, ethyl 2-ethylamino ethyl amine, ethylenediamine, n,n'-diethyl, n,n'-diethyl-1,2-diaminoethane, dihydrobromide | |
CJKRXEBLWJVYJD-UHFFFAOYSA-N | |
N,N'-diethylethane-1,2-diamine | |
67105 | |
116.21 | |
N, N'-Diethylethylenediamine |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-902-4
- N,N'-Diethylethylenediamine
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.