missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diformylacetamide, 80%, Thermo Scientific Chemicals

Product Code. 10679404
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26944-31-0
Molecular Formula C4H5NO3
Molecular Weight (g/mol) 115.09
MDL Number MFCD00015916
InChI Key ZAHBLOGBMDVRDK-UHFFFAOYSA-N
Synonym n,n-diformylacetamid
PubChem CID 8063338
IUPAC Name N,N-diformylacetamide
SMILES CC(=O)N(C=O)C=O

Specifications

CAS Min % 75.0
CAS Max % 100.0
Color Colorless
Boiling Point 70°C (15.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 75% min. (GC)
Packaging Glass bottle
Linear Formula CH3C(O)N(COH)2
Quantity 5 g
Formula Weight 115.09
Percent Purity 80%
Physical Form Liquid
Chemical Name or Material N, N-Diformylacetamide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'