missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Diisopropylethylenediamine, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 mL
100 mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Density | 0.8300g/mL |
Boiling Point | 170°C |
Flash Point | 54°C |
Assay Percent Range | 97% |
Packaging | Glass bottle |
Linear Formula | [(CH3)2CH]2NCH2CH2NH2 |
Quantity | 100 mL |
Specific Gravity | 0.83 |
Solubility Information | Solubility in water: miscible |
Formula Weight | 144.26 |
Show More |
Safety and Handling
Product Identifier
- N,N-Diisopropylethylenediamine
- Danger
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction