missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-1,4-butanediamine, 98%, Thermo Scientific Chemicals

Product Code. 11449410
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

N,N-Dimethyl-1,4-butanediamine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethyl-1,4-butanediamine is used as pharmaceutical intermediate.

Notes
Air sensitive. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3529-10-0
Molecular Formula C6H16N2
Molecular Weight (g/mol) 116.208
MDL Number MFCD00047977
InChI Key GCOWZPRIMFGIDQ-UHFFFAOYSA-N
Synonym 4-dimethylaminobutylamine, 4-aminobutyl dimethylamine, 4-dimethylamino butylamine, n,n-dimethyl-1,4-butanediamine, n,n-dimethylbutane-1,4-diamine, n,n-dimethylaminobutylamine, chembl59625, nn-dimethyl-1,4-diaminobutane, 1,4-butanediamine, n,n-dimethyl, n1,n1-dimethyl-1,4-butanediamine
PubChem CID 77063
IUPAC Name N',N'-dimethylbutane-1,4-diamine
SMILES CN(C)CCCCN

Specifications

Linear Formula (CH3)2NCH2CH2CH2CH2NH2
Refractive Index 1.44
Quantity 5 g
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 116.21
Percent Purity 98%
Chemical Name or Material N,N-Dimethyl-1,4-butanediamine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'