Learn More
N,N-Dimethyl-1-naphthylamine, 99%, Thermo Scientific Chemicals
Description
N,N-Dimethyl-1-naphthylamine is used in the nitrate reductase test to form a red precipitate of Prontosil by reacting with a nitrite-sulfanilic acid complex. And is also used to detection and determination of nitrites.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Yellow |
Density | 1.039 g/mL |
Boiling Point | 145°C to 147°C (12 mmHg) |
Refractive Index | 1.623 |
Quantity | 5 mL |
UN Number | UN3082 |
Merck Index | 14,3252 |
Solubility Information | Insoluble in water. |
Formula Weight | 171.24 |
Percent Purity | 99% |
Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.