missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-1-naphthylamine, 99%, Thermo Scientific Chemicals

Product Code. 10723534
Click to view available options
Quantity:
5 mL
25 mL
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-56-6
Molecular Formula C12H13N
Molecular Weight (g/mol) 171.24
MDL Number MFCD00003919
InChI Key AJUXDFHPVZQOGF-UHFFFAOYSA-N
Synonym n,n-dimethyl-1-naphthylamine, 1-dimethylaminonaphthalene, 1-naphthalenamine, n,n-dimethyl, n,n-dimethyl-1-naphthalenamine, n,n-dimethyl-1-napthylamine, dimethyl 1-naphthyl amine, alpha-dimethylaminonaphthalene, 1-naphthylamine, n,n-dimethyl, dimethyl-alpha-naphthylamine, n,n-dimethyl-1-naftylamin
PubChem CID 6848
IUPAC Name N,N-dimethylnaphthalen-1-amine
SMILES CN(C)C1=CC=CC2=CC=CC=C21

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Yellow to Brown
Density 1.0400g/mL
Boiling Point 139°C to 140°C (13.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C10H7N(CH3)2
Refractive Index 1.6210 to 1.623
Quantity 500 mL
Beilstein 12, 1221
Merck Index 15, 3276
Specific Gravity 1.04
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 171.24
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N, N-Dimethyl-1-naphthylamine, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N,N-Dimethyl-1-naphthylamine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'