Learn More
N,N-Dimethyl-4-nitrosoaniline, 98%, Thermo Scientific Chemicals
32.66 € - 184.92 €
Chemical Identifiers
CAS | 138-89-6 |
---|---|
Molecular Formula | C8H10N2O |
Molecular Weight (g/mol) | 150.181 |
MDL Number | MFCD00002063 |
InChI Key | CMEWLCATCRTSGF-UHFFFAOYSA-N |
Synonym | ndma, accelerine, p-nitroso-n,n-dimethylaniline, n,n-dimethyl-p-nitrosoaniline, 4-nitroso-n,n-dimethylaniline, 4-nitrosodimethylaniline, p-nitrosodimethylanilide, paranitrosodimethylanilide, 4-dimethylamino nitrosobenzene, benzenamine, n,n-dimethyl-4-nitroso |
PubChem CID | 8749 |
ChEBI | CHEBI:59990 |
IUPAC Name | N,N-dimethyl-4-nitrosoaniline |
SMILES | CN(C)C1=CC=C(C=C1)N=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11453847
|
Thermo Scientific Alfa Aesar
A18301.14 |
25 g |
32.66 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11463847
|
Thermo Scientific Alfa Aesar
A18301.22 |
100 g |
91.56 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11498770
|
Thermo Scientific Alfa Aesar
A18301.30 |
250 g |
184.92 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
138-89-6 | |
150.181 | |
CMEWLCATCRTSGF-UHFFFAOYSA-N | |
8749 | |
N,N-dimethyl-4-nitrosoaniline |
C8H10N2O | |
MFCD00002063 | |
ndma, accelerine, p-nitroso-n,n-dimethylaniline, n,n-dimethyl-p-nitrosoaniline, 4-nitroso-n,n-dimethylaniline, 4-nitrosodimethylaniline, p-nitrosodimethylanilide, paranitrosodimethylanilide, 4-dimethylamino nitrosobenzene, benzenamine, n,n-dimethyl-4-nitroso | |
CHEBI:59990 | |
CN(C)C1=CC=C(C=C1)N=O |
Specifications
138-89-6 | |
1.15 | |
MFCD00002063 | |
607293 | |
ndma, accelerine, p-nitroso-n,n-dimethylaniline, n,n-dimethyl-p-nitrosoaniline, 4-nitroso-n,n-dimethylaniline, 4-nitrosodimethylaniline, p-nitrosodimethylanilide, paranitrosodimethylanilide, 4-dimethylamino nitrosobenzene, benzenamine, n,n-dimethyl-4-nitroso | |
CN(C)C1=CC=C(C=C1)N=O | |
150.181 | |
CHEBI:59990 | |
98% |
84°C to 87°C | |
C8H10N2O | |
UN1369 | |
14,6638 | |
CMEWLCATCRTSGF-UHFFFAOYSA-N | |
N,N-dimethyl-4-nitrosoaniline | |
8749 | |
150.18 | |
N,N-Dimethyl-4-nitrosoaniline |
Safety and Handling
GHS H Statement
H228-H251-H301-H310-H351-H373
Flammable solid.
Self-heating: may catch fire.
Toxic if swallowed.
Fatal in contact with skin.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
P210-P235+P410-P260-P262-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P362-P370+P380-P380-P501c
H301+H311+H331-H373-H500-X
missing translation for 'dotInformation' : Transport Hazard Class: 4.2; Packing Group: II; Proper Shipping Name: P-NITROSODIMETHYLANILINE
missing translation for 'einecsNumber' : 205-343-1
missing translation for 'rtecsNumber' : BX7175000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only