missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-m-phenylenediamine dihydrochloride, 99%, Thermo Scientific Chemicals

58.69 € - 547.00 €

Chemical Identifiers

CAS 3575-32-4
Molecular Formula C8H12N2·2HCl
Molecular Weight (g/mol) 209.12
MDL Number MFCD00012971
InChI Key BZJPIQKDEGXVFG-UHFFFAOYSA-N
Synonym n1,n1-dimethylbenzene-1,3-diamine dihydrochloride, n,n-dimethyl-m-phenylenediamine dihydrochloride, m-dimethylamino aniline, n,n-dimethyl-1,3-phenylenediamine dihydrochloride, 1,3-benzenediamine, n,n-dimethyl-, dihydrochloride, n,n-dimethylbenzene-1,3-diamine dihydrochloride, 3-amino-n,n-dimethylaniline, 1,3-benzenediamine, n1,n1-dimethyl-, hydrochloride 1:2, pubchem14991, acmc-20a5k9
PubChem CID 77124
IUPAC Name 3-N,3-N-dimethylbenzene-1,3-diamine;dihydrochloride
SMILES CN(C)C1=CC=CC(=C1)N.Cl.Cl
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10791091
View Documents
Thermo Scientific Acros
204340050
5 g Glass bottle
58.69 €
5g
Please to purchase this item. Need a web account? Register with us today!
10368050
View Documents
Thermo Scientific
204340250
25 g Glass bottle
191.96 €
25g
Please to purchase this item. Need a web account? Register with us today!
10351514
View Documents
Thermo Scientific Acros
204341000
100 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3575-32-4
209.12
BZJPIQKDEGXVFG-UHFFFAOYSA-N
77124
CN(C)C1=CC=CC(=C1)N.Cl.Cl
C8H12N2·2HCl
MFCD00012971
n1,n1-dimethylbenzene-1,3-diamine dihydrochloride, n,n-dimethyl-m-phenylenediamine dihydrochloride, m-dimethylamino aniline, n,n-dimethyl-1,3-phenylenediamine dihydrochloride, 1,3-benzenediamine, n,n-dimethyl-, dihydrochloride, n,n-dimethylbenzene-1,3-diamine dihydrochloride, 3-amino-n,n-dimethylaniline, 1,3-benzenediamine, n1,n1-dimethyl-, hydrochloride 1:2, pubchem14991, acmc-20a5k9
3-N,3-N-dimethylbenzene-1,3-diamine;dihydrochloride

Specifications

3575-32-4
100.0
Purple to White
98.5% min. (Argentometry)
(CH3)2NC6H4NH2·2HCl
13, 40
Solubility in water: soluble
CN(C)C1=CC=CC(=C1)N.Cl.Cl
209.12
209.12
Crystalline Powder
98.5
217.0°C
Authentic
C8H12N2·2HCl
MFCD00012971
n1,n1-dimethylbenzene-1,3-diamine dihydrochloride, n,n-dimethyl-m-phenylenediamine dihydrochloride, m-dimethylamino aniline, n,n-dimethyl-1,3-phenylenediamine dihydrochloride, 1,3-benzenediamine, n,n-dimethyl-, dihydrochloride, n,n-dimethylbenzene-1,3-diamine dihydrochloride, 3-amino-n,n-dimethylaniline, 1,3-benzenediamine, n1,n1-dimethyl-, hydrochloride 1:2, pubchem14991, acmc-20a5k9
BZJPIQKDEGXVFG-UHFFFAOYSA-N
3-N,3-N-dimethylbenzene-1,3-diamine;dihydrochloride
77124
99%
N, N-Dimethyl-m-phenylenediamine dihydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
Toxic if inhaled.
Causes serious eye irritation.
Toxic if swallowed.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 222-693-

Product Identifier
  • N,N-Dimethyl-m-phenylenediamine dihydrochloride

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.