missing translation for 'onlineSavingsMsg'
Learn More
N,N-Dimethyl-n-octylamine, 95%, Thermo Scientific Chemicals Product Code.: 11454234

N,N-Dimethyl-n-octylamine, 95%, Thermo Scientific Chemicals

Product Code. 11454234
10 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11454234

Brand: Thermo Scientific Alfa Aesar A12546.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

N,N-Dimethyl-n-octylamine is used as ion-pairing agent in the quantitative estimation of thiazinamium methylsulphate in a pharmaceutical preparation by HPLC. It acts as a counterion in mobile phase for separation of tetracycline, tetracycline analogs and their potential impurities by reversed-phase ion-pair chromatography. Further, it is used in the study of chiral separations by complexation with proteins in capillary zone electrophoresis. In addition to this, it is used as a mobile phase in direct resolution and quantitation of (R)- and (S)-disopyramide

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethyl-n-octylamine is used as ion-pairing agent in the quantitative estimation of thiazinamium methylsulphate in a pharmaceutical preparation by HPLC. It acts as a counterion in mobile phase for separation of tetracycline, tetracycline analogs and their potential impurities by reversed-phase ion-pair chromatography. Further, it is used in the study of chiral separations by complexation with proteins in capillary zone electrophoresis. In addition to this, it is used as a mobile phase in direct resolution and quantitation of (R)- and (S)-disopyramide

Solubility
Slightly miscible with water.

Notes
Air sensitive. Incompatible with strong oxidizing agents, strong acids and copper.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7378-99-6
Molecular Formula C10H23N
Molecular Weight (g/mol) 157.30
MDL Number MFCD00009558
InChI Key UQKAOOAFEFCDGT-UHFFFAOYSA-N
Synonym n,n-dimethyloctylamine, n,n-dimethyl-n-octylamine, dimethyloctylamine, octyldimethylamine, dimethyl octyl amine, n-octyldimethylamine, dimethyl-n-octylamine, 1-octanamine, n,n-dimethyl, octylamine, n,n-dimethyl, unii-20n7h7x4sd
PubChem CID 16224
IUPAC Name N,N-dimethyloctan-1-amine
SMILES CCCCCCCCN(C)C

Specifications

Melting Point -57°C
Density 0.765
Boiling Point 195°C
Flash Point 65°C (149°F)
Refractive Index 1.4243
Quantity 10 g
UN Number UN2922
Beilstein 1698081
Sensitivity Air Sensitive
Solubility Information Slightly miscible with water.
Formula Weight 157.3
Percent Purity 95%
Chemical Name or Material N,N-Dimethyl-n-octylamine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0456675 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA12546.18,A12546.09,A12546.30
Lot NumberA0455550 Certificate TypeCertificate of Analysis Date19/10/2023 Catalog NumberA12546.18,A12546.09,A12546.30
Lot NumberA0434757 Certificate TypeCertificate of Analysis Date19/10/2022 Catalog NumberA12546.18,A12546.09,A12546.30
Lot Number10130736 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12546.18,A12546.09,A12546.30
Lot NumberE19Q25 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12546.18,A12546.09,A12546.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N,N-Dimethyl-n-octylamine, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.