Learn More
N,N-Dimethyl-p-phenylenediamine, 97%, Thermo Scientific Chemicals
37.99 € - 329.64 €
Chemical Identifiers
CAS | 99-98-9 |
---|---|
Molecular Formula | C8H12N2 |
Molecular Weight (g/mol) | 136.20 |
MDL Number | MFCD00007860 |
InChI Key | BZORFPDSXLZWJF-UHFFFAOYSA-N |
Synonym | n,n-dimethyl-p-phenylenediamine, n,n-dimethyl-1,4-phenylenediamine, n1,n1-dimethylbenzene-1,4-diamine, p-aminodimethylaniline, dmpd, 4-dimethylamino aniline, dimethyl-p-phenylenediamine, p-amino-n,n-dimethylaniline, 4-amino-n,n-dimethylaniline, p-dimethylamino aniline |
PubChem CID | 7472 |
ChEBI | CHEBI:15783 |
IUPAC Name | 4-N,4-N-dimethylbenzene-1,4-diamine |
SMILES | CN(C)C1=CC=C(N)C=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10137470
|
Thermo Scientific Acros
408460050 |
5 g | Glass bottle |
37.99 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11310976
|
Thermo Scientific Acros
408460250 |
25 g | Glass bottle |
37.99 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10219343
|
Thermo Scientific Acros
408461000 |
100 g | Glass bottle |
92.76 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10772865
|
Thermo Scientific Acros
408465000 |
500 g | Glass bottle |
329.64 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
99-98-9 | |
136.20 | |
BZORFPDSXLZWJF-UHFFFAOYSA-N | |
7472 | |
4-N,4-N-dimethylbenzene-1,4-diamine |
C8H12N2 | |
MFCD00007860 | |
n,n-dimethyl-p-phenylenediamine, n,n-dimethyl-1,4-phenylenediamine, n1,n1-dimethylbenzene-1,4-diamine, p-aminodimethylaniline, dmpd, 4-dimethylamino aniline, dimethyl-p-phenylenediamine, p-amino-n,n-dimethylaniline, 4-amino-n,n-dimethylaniline, p-dimethylamino aniline | |
CHEBI:15783 | |
CN(C)C1=CC=C(N)C=C1 |
Specifications
99-98-9 | |
100.0 | |
Brown | |
262.0°C | |
Authentic | |
C8H12N2 | |
MFCD00007860 | |
1.09 | |
Solubility in water: 11g/L (20°C). Other solubilities: soluble in alcohol,chloroform,ether,benzene | |
CN(C)C1=CC=C(N)C=C1 | |
136.20 | |
CHEBI:15783 | |
97% | |
N, N-Dimethyl-p-phenylenediamine, Practical |
96.0 | |
35.0°C | |
1.0900g/mL | |
130°C | |
96% min. (GC) | |
NH2C6H4N(CH3)2 | |
15, 3278 | |
n,n-dimethyl-p-phenylenediamine, n,n-dimethyl-1,4-phenylenediamine, n1,n1-dimethylbenzene-1,4-diamine, p-aminodimethylaniline, dmpd, 4-dimethylamino aniline, dimethyl-p-phenylenediamine, p-amino-n,n-dimethylaniline, 4-amino-n,n-dimethylaniline, p-dimethylamino aniline | |
BZORFPDSXLZWJF-UHFFFAOYSA-N | |
4-N,4-N-dimethylbenzene-1,4-diamine | |
7472 | |
136.2 | |
Crystalline Low Melting Solid |
Safety and Handling
GHS H Statement
Fatal if swallowed.
Toxic if inhaled.
Toxic in contact with skin.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 202-807-5
- N,N-Dimethyl-p-phenylenediamine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 3
Hazard Statement
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H300-Fatal if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P330-Rinse mouth.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.