missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethylacetamide, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Packaging:
AcroSeal™ Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | -20.0°C |
CAS Min % | 99.4 |
Color | Colorless |
CAS Max % | 100.0 |
Density | 0.9370g/mL |
Boiling Point | 164.0°C to 166.0°C |
Quantity | 1 L |
Flash Point | 70°C |
Infrared Spectrum | Authentic |
Packaging | AcroSeal™ Glass Bottle |
Show More |
Safety and Handling


Product Identifier
- N,N-Dimethylacetamide
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H360D-May damage the unborn child.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
N,N-Dimethylacetamide, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction