missing translation for 'onlineSavingsMsg'
Learn More
N,N-Dimethylacetamide, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals Product Code.: 10023743

N,N-Dimethylacetamide, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10023743
100 mL, AcroSeal™ Glass Bottle
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

Product Code. 10023743

Brand: Thermo Scientific Acros 375231000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-19-5
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00008686
InChI Key FXHOOIRPVKKKFG-UHFFFAOYSA-N
Synonym dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamineShow MoreShow Less
PubChem CID 31374
ChEBI CHEBI:84254
IUPAC Name N,N-dimethylacetamide
SMILES CN(C)C(C)=O

Specifications

CAS Min % 99.4
CAS Max % 100.0
Melting Point -20.0°C
Color Colorless
Density 0.9370g/mL
Boiling Point 164.0°C to 166.0°C
Flash Point 70°C
Infrared Spectrum Authentic
Assay Percent Range 99.4% min. (GC)
Packaging AcroSeal™ Glass Bottle
Linear Formula CH3CON(CH3)2
Refractive Index 1.4370 to 1.4390
Quantity 100 mL
Merck Index 15, 3248
Specific Gravity 0.937
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acetone,diethyl ether,benzene,,chloroform,ethanol and toluene
Viscosity 1.02 mPa.s (20°C)
Formula Weight 87.12
Percent Purity 99.5%
Grade Water <50ppm
Physical Form Liquid
Chemical Name or Material N, N-Dimethylacetamide, Over Molecular Sieves
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2497440 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number375231000,375230010
Lot Number2413371 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number375231000,375230010
Lot Number2418109 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number375231000,375230010
Lot Number1911062 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number375231000,375230010
Lot Number1905940 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number375231000,375230010
compliance-icons compliance-icons
Product Identifier
  • N,N-Dimethylacetamide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : dimethyl acetamide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N,N-Dimethylacetamide, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.