missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylbenzylamine, 99%

Product Code. 10531411 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
5 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
100mL
2.5 L
5 L
500mL
This item is not returnable. View return policy

Product Code. 10531411

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 103-83-3 | C9H13N | 135.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-83-3
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00008329
InChI Key XXBDWLFCJWSEKW-UHFFFAOYSA-N
Synonym n,n-dimethylbenzylamine, benzyldimethylamine, n-benzyldimethylamine, dimethylbenzylamine, bdma, benzenemethanamine, n,n-dimethyl, benzyl-n,n-dimethylamine, n-phenylmethyl dimethylamine, n,n-dimethylbenzenemethanamine, araldite accelerator 062
PubChem CID 7681
SMILES CN(C)CC1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -75.0°C
Color Colorless to Yellow
Density 0.9000g/mL
Boiling Point 183.0°C to 184.0°C
Flash Point 54°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2N(CH3)2
Refractive Index 1.5000 to 1.5020
Quantity 500 mL
Beilstein 12, 1019
Specific Gravity 0.9
Solubility Information Solubility in water: 8g/L (20°C). Other solubilities: soluble in alcohol,acetone,ether and toluene,reacts with organic and inorganic acids
IUPAC Name N,N-dimethyl-1-phenylmethanamine
Formula Weight 135.23
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N, N-Dimethylbenzylamine, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N,N-Dimethylbenzylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.