missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethylformamide, 99.5%, for analysis, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mL
1 L
2.5 L
10 L
25 L
Packaging:
Glass Bottle
Plastic Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis

Specifications
Specifications
Melting Point | -61.0°C |
CAS Min % | 99.4 |
Color | Colorless |
CAS Max % | 100.0 |
Density | 0.9450g/mL |
Boiling Point | 153.0°C |
Quantity | 2.5 L |
Flash Point | 58°C |
Infrared Spectrum | Authentic |
Packaging | Glass Bottle |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2414286 | Certificate of Analysis | 31/01/2025 | 210580025,210580100,210580010,210585000,210580250 |
2402191 | Certificate of Analysis | 31/01/2025 | 210580025,210580100,210580010,210585000,210580250 |
2420924 | Certificate of Analysis | 31/01/2025 | 210580025,210580100,210580010,210585000,210580250 |
2401363 | Certificate of Analysis | 31/01/2025 | 210580025,210580100,210580010,210585000,210580250 |
2404045 | Certificate of Analysis | 31/01/2025 | 210580025,210580100,210580010,210585000,210580250 |
Safety and Handling



Product Identifier
- N,N-Dimethylformamide
- Danger
- Flammable liquid Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
- H226-Flammable liquid and vapour.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H360D-May damage the unborn child.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
N,N-Dimethylformamide, 99.5%, for analysis, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction