missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide, ≥99.8%, ACS Reagent, Honeywell Riedel-de Haën™

Product Code. 15683600
Click to view available options
Quantity:
500 mL
1 L
2.5 L
20 L
Packaging:
Glass Bottle
Stainless Steel Can
This item is not returnable. View return policy
This item is not returnable. View return policy

ACS Reagent,≥99.8%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

Melting Point -61°C
Density 0.944g/mL
Boiling Point 153°C
Quantity 2.5 L
Explosion Limit 0.152
Flash Point 58°C (136.4°F)
Ignition Point 833°F
Packaging Glass Bottle
Linear Formula HCON(CH3)2
Vapor Pressure 2.7mmHg (20°C)
UN Number UN2265
Beilstein 605365
Formula Weight 73.09g/mol
Percent Purity ≥99.8%
Grade ACS Reagent
Chemical Name or Material N,N-Dimethylformamide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'