missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethylformamide, ≥99.8%, ACS Reagent, Honeywell Riedel-de Haën™
ACS Reagent,≥99.8%
45.49 € - 993.13 €
Chemical Identifiers
CAS | 68-12-2 |
---|---|
Molecular Formula | C3H7NO |
Molecular Weight (g/mol) | 73.10 |
MDL Number | MFCD00003284 |
InChI Key | ZMXDDKWLCZADIW-UHFFFAOYSA-N |
Synonym | dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa |
PubChem CID | 6228 |
ChEBI | CHEBI:17741 |
IUPAC Name | N,N-dimethylformamide |
SMILES | CN(C)C=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15603610
|
Honeywell-Riedel-de Haen
319937-500ML |
500 mL | Glass Bottle | ||||||
15673600
|
Honeywell-Riedel-de Haen
319937-1L |
1 L | Glass Bottle | ||||||
15683600
|
Honeywell-Riedel-de Haen
319937-2.5L |
2.5 L | Glass Bottle | ||||||
15693600
|
Honeywell-Riedel-de Haen
319937-20L |
20 L | Stainless Steel Can | ||||||
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Chemical Identifiers
68-12-2 | |
73.10 | |
ZMXDDKWLCZADIW-UHFFFAOYSA-N | |
6228 | |
N,N-dimethylformamide |
C3H7NO | |
MFCD00003284 | |
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa | |
CHEBI:17741 | |
CN(C)C=O |
Specifications
-61°C | |
0.944g/mL | |
0.152 | |
833°F | |
HCON(CH3)2 | |
2.7mmHg (20°C) | |
605365 | |
ZMXDDKWLCZADIW-UHFFFAOYSA-N | |
N,N-dimethylformamide | |
6228 | |
73.09g/mol | |
ACS Reagent |
68-12-2 | |
153°C | |
58°C (136.4°F) | |
C3H7NO | |
MFCD00003284 | |
UN2265 | |
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa | |
CN(C)C=O | |
73.10 | |
CHEBI:17741 | |
≥99.8% | |
N,N-Dimethylformamide |
Safety and Handling
P201-P210-P261-P280-P308 + P313-P370 + P378
H226-H312 + H332-H319-H360D
missing translation for 'dotInformation' : 3/PGIII
missing translation for 'einecsNumber' : 200-679-5
missing translation for 'rtecsNumber' : LQ2100000