missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethylformamide, ≥99.8%, ACS reagent
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
500 mL
1 L
2.5 L
10 L
Packaging:
EcoSafPak™ Glass Bottle
Glass Bottle
Plastic Drum
Unit Size:
1 L
10 L
2.5 L
25mL
500mL
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Especificaciones
| Melting Point | -61.0°C |
| CAS Min % | 99.8 |
| CAS Max % | 100.0 |
| Decomposition Information | >350°C |
| Density | 0.9450g/mL |
| Boiling Point | 153.0°C |
| Quantity | 25 mL |
| Flash Point | 58°C |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Mostrar más |
Seguridad y manipulación
Nombre del producto
- N,N-Dimethylformamide
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Reproductive toxicity Category 1B
- H226-Flammable liquid and vapour.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H360D-May damage the unborn child.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : N,N-Dimethylformamide
RUO – Research Use Only
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido