missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide-d7, for NMR, packaged in 0.75 ml ampoules, 99.5 atom % D, Thermo Scientific Chemicals

Product Code. 10389170
Click to view available options
Quantity:
7.5 mL
Packaging:
Ampule
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Specifications

Physical Form Liquid
Color Colorless
Density 1.0300g/mL
Deuterium (D) 99.4% min.
Flash Point 58°C
Packaging Ampule
Refractive Index 1.428
Water 0.1% max. (Coulometric)
Quantity 7.5 mL
CAS 4472-41-7
Molecular Formula C3H7NO
Linear Formula DCON(CD3)2
MDL Number MFCD00003286
Synonym n,n-dimethylformamide-d7, dmf-d7, deuterated dmf, n,n-di 2h3 methyl 2h formamide, dimethyl formamide-d7, dimethylformamide d, heptadeutero-n,n-dimethylformamide, n,n-dimethylformamide-d7 dmf-d7, formamide-1-d, n,n-di methyl-d3, n,n-dimethylformamide-d7 99.5atom%d
InChI Key ZMXDDKWLCZADIW-YYWVXINBSA-N
SMILES CN(C)C=O
IUPAC Name 1-deuterio-N,N-bis(trideuteriomethyl)formamide
Molecular Weight (g/mol) 80.138
PubChem CID 78225
Formula Weight 80.15
Grade NMR
Chemical Name or Material N, N-Dimethylformamide-d7
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N,N-Dimethylformamide-d7
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1A
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H319-Causes serious eye irritation.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'