missing translation for 'onlineSavingsMsg'
Learn More
N,N-Dimethylformamide dibenzyl acetal, tech. 80%, Thermo Scientific Chemicals Product Code.: 11453644

N,N-Dimethylformamide dibenzyl acetal, tech. 80%, Thermo Scientific Chemicals

Product Code. 11453644
250 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11453644

Brand: Thermo Scientific Alfa Aesar A11281.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

N,N-Dimethylformamide diethyl acetal was used in quantification of cocaine and its primary metabolite, benzoyl ecgonine from urine matrix by gas chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylformamide diethyl acetal was used in quantification of cocaine and its primary metabolite, benzoyl ecgonine from urine matrix by gas chromatography.

Solubility
Soluble in variety of inert solvents.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2016-04-8
Molecular Formula C17H22NO2
Molecular Weight (g/mol) 272.37
MDL Number MFCD00014869
InChI Key JFIKHFNGAURIIB-UHFFFAOYSA-O
Synonym n,n-dimethylformamide dibenzyl acetal, n,n-dimethylformamide dibenzylacetal, 1,1-bis benzyloxy trimethylamine, methanamine, n,n-dimethyl-1,1-bis phenylmethoxy, bis benzyloxy methyl dimethylamine, acmc-1cro2, 1,1-dibenzyloxytrimethylamine, dimethyl formamide dibenzylacetal, bis benzyloxy methyldimethylamine, dimethyl formamide dibenzyl acetal
PubChem CID 74837
IUPAC Name N,N-dimethyl-1,1-bis(phenylmethoxy)methanamine
SMILES CN(C)C(OCC1=CC=CC=C1)OCC2=CC=CC=C2

Specifications

Density 1.045
Boiling Point 138°C to 140°C (0.5 mmHg)
Quantity 250 g
Sensitivity Moisture sensitive
Solubility Information Soluble in variety of inert solvents.
Formula Weight 271.36
Percent Purity 85%
Grade Technical
Chemical Name or Material N,N-Dimethylformamide dibenzyl acetal

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N,N-Dimethylformamide dibenzyl acetal, tech. 80%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.